Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AFP
Entrez Gene ID
174
Gene Name
alpha fetoprotein
Gene Aliases
AFPD, FETA, HPAFP
Chromosome Location
Chr.4: 73436219 - 73455785 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134.2 NP_001125.1 12 - 13 62
GenBank mRNA AI336986.1 - 3 - 4 361 62
AK297669.1 - 12 - 13 62
AK297674.1 - 12 - 13 62
AK314817.1 - 12 - 13 62
BC027881.1 - 12 - 13 62
V01514.1 - 12 - 13 62

View Full Product Details