The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001134.2 | NP_001125.1 | 2 - 3 | 175 | 78 |
GenBank mRNA | AK297669.1 | - | 2 - 3 | 173 | 78 |
AK297674.1 | - | 2 - 3 | 170 | 78 | |
AK314817.1 | - | 2 - 3 | 169 | 78 | |
BC027881.1 | - | 2 - 3 | 161 | 78 | |
V01514.1 | - | 2 - 3 | 175 | 78 |