The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_014239.3 | NP_055054.1 | 2 - 3 | 363 | 80 |
GenBank mRNA | AF035280.1 | - | 2 - 3 | 327 | 80 |
BC000494.2 | - | 2 - 3 | 329 | 80 | |
BC003165.1 | - | 2 - 3 | 329 | 80 | |
BC011750.2 | - | 2 - 3 | 304 | 80 | |
BI908906.1 | - | 2 - 3 | 363 | 80 | |
BX161473.1 | - | 2 - 3 | 361 | 80 |