Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
UPK1B
Entrez Gene ID
7348
Gene Name
uroplakin 1B
Gene Aliases
TSPAN20, UPIB, UPK1
Chromosome Location
Chr.3: 119173578 - 119205153 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_006952.3 NP_008883.2 2 - 3 161 75
GenBank mRNA AB002155.1 - 2 - 3 119 75
AB015234.1 - 1 - 2 72 75
AF042331.1 - 1 - 2 82 75
AF082888.1 - 1 - 2 42 75
AK225868.1 - 1 - 2 76 75
AK313787.1 - 2 - 3 183 75
AL709739.1 - 2 - 3 162 75
BC063568.1 - 1 - 2 109 75
CR457084.1 - 1 - 2 72 75

View Full Product Details