The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145138.1 | NP_001138610.1 | 2 - 3 | 170 | 83 |
NM_001243984.1 | NP_001230913.1 | 2 - 3 | 170 | 83 | |
NM_001243985.1 | NP_001230914.1 | 2 - 3 | 170 | 83 | |
NM_021975.3 | NP_068810.3 | 2 - 3 | 170 | 83 | |
XM_011545206.1 | XP_011543508.1 | 2 - 3 | 151 | 83 | |
XM_011545207.1 | XP_011543509.1 | 2 - 3 | 151 | 83 | |
GenBank mRNA | AK303262.1 | - | 2 - 3 | 116 | 83 |
AK308020.1 | - | 2 - 3 | 114 | 83 | |
AK308478.1 | - | 2 - 3 | 115 | 83 | |
BC011603.1 | - | 2 - 3 | 93 | 83 | |
BC014095.2 | - | 2 - 3 | 102 | 83 | |
BC033522.1 | - | 1 - 2 | 36 | 83 | |
BC110830.1 | - | 2 - 3 | 93 | 83 | |
BM916512.1 | - | 2 - 3 | 91 | 83 | |
DA020095.1 | - | 2 - 3 | 292 | 83 | |
M62399.1 | - | 2 - 3 | 109 | 83 |