Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CCDC77
Entrez Gene ID
84318
Gene Name
coiled-coil domain containing 77
Gene Aliases
-
Chromosome Location
Chr.12: 389350 - 442642 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001130146.1 NP_001123618.1 11 - 12 1451 78
NM_001130147.1 NP_001123619.1 11 - 12 1478 78
NM_001130148.1 NP_001123620.1 10 - 11 1424 78
NM_032358.3 NP_115734.1 12 - 13 1505 78
GenBank mRNA AK027638.1 - 12 - 13 1423 78
AK293165.1 - 11 - 12 1389 78
AK316029.1 - 10 - 11 1426 78
BC006444.2 - 11 - 12 1394 78

View Full Product Details