Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
C8orf44
Entrez Gene ID
56260
Gene Name
chromosome 8 open reading frame 44
Gene Aliases
-
Chromosome Location
Chr.8: 66667552 - 66681149 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK002129.1 - 1 - 2 69 74

View Full Product Details