Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
DNAH2
Entrez Gene ID
146754
Gene Name
dynein axonemal heavy chain 2
Gene Aliases
DNAHC2, DNHD3
Chromosome Location
Chr.17: 7717354 - 7833744 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_020877.3 NP_065928.2 20 - 21 4802 84
XM_011523663.1 XP_011521965.1 22 - 23 3670 84
XM_011523664.2 XP_011521966.1 23 - 24 3819 84
XM_011523666.1 XP_011521968.1 22 - 23 3670 84
XM_011523667.2 XP_011521969.1 21 - 22 3573 84
XM_011523668.1 XP_011521970.1 12 - 13 2053 84
XM_011523669.2 XP_011521971.1 10 - 11 1737 84
XM_011523670.2 XP_011521972.1 22 - 23 3670 84
XM_017024218.1 XP_016879707.1 9 - 10 1754 84
XM_017024219.1 XP_016879708.1 22 - 23 3670 84
GenBank mRNA AB040936.2 - 20 - 21 4802 84
BX648163.1 - 3 - 4 556 84

View Full Product Details