The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020877.3 | NP_065928.2 | 25 - 26 | 5561 | 82 |
XM_011523663.1 | XP_011521965.1 | 27 - 28 | 4429 | 82 | |
XM_011523664.2 | XP_011521966.1 | 28 - 29 | 4578 | 82 | |
XM_011523666.1 | XP_011521968.1 | 27 - 28 | 4429 | 82 | |
XM_011523667.2 | XP_011521969.1 | 26 - 27 | 4332 | 82 | |
XM_011523668.1 | XP_011521970.1 | 17 - 18 | 2812 | 82 | |
XM_011523669.2 | XP_011521971.1 | 15 - 16 | 2496 | 82 | |
XM_011523670.2 | XP_011521972.1 | 27 - 28 | 4429 | 82 | |
XM_017024218.1 | XP_016879707.1 | 14 - 15 | 2513 | 82 | |
XM_017024219.1 | XP_016879708.1 | 27 - 28 | 4429 | 82 | |
GenBank mRNA | AB040936.2 | - | 25 - 26 | 5561 | 82 |
BX648163.1 | - | 7 - 8 | 1151 | 82 |