The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_194318.3 | NP_919299.3 | 10 - 11 | 959 | 101 |
XM_006719768.3 | XP_006719831.1 | 11 - 12 | 1149 | 101 | |
XM_011534936.1 | XP_011533238.1 | 10 - 11 | 948 | 101 | |
XM_011534938.2 | XP_011533240.1 | 9 - 10 | 797 | 101 | |
XM_017020395.1 | XP_016875884.1 | 11 - 12 | 1052 | 101 | |
GenBank mRNA | AB101481.1 | - | 10 - 11 | 849 | 101 |
AK291273.1 | - | 10 - 11 | 948 | 101 | |
AY190526.1 | - | 10 - 11 | 849 | 101 | |
DA290408.1 | - | 3 - 4 | 256 | 101 |