The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_194318.3 | NP_919299.3 | 11 - 12 | 1070 | 96 |
XM_006719768.3 | XP_006719831.1 | 12 - 13 | 1260 | 96 | |
XM_011534936.1 | XP_011533238.1 | 11 - 12 | 1059 | 96 | |
XM_011534938.2 | XP_011533240.1 | 10 - 11 | 908 | 96 | |
XM_017020395.1 | XP_016875884.1 | 12 - 13 | 1163 | 96 | |
GenBank mRNA | AB101481.1 | - | 11 - 12 | 960 | 96 |
AK291273.1 | - | 11 - 12 | 1059 | 96 | |
AY190526.1 | - | 11 - 12 | 960 | 96 | |
BC068595.1 | - | 11 - 12 | 1050 | 96 | |
DA290408.1 | - | 4 - 5 | 367 | 96 |