The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_194318.3 | NP_919299.3 | 7 - 8 | 699 | 130 |
XM_006719768.3 | XP_006719831.1 | 8 - 9 | 889 | 130 | |
XM_011534936.1 | XP_011533238.1 | 7 - 8 | 688 | 130 | |
XM_011534938.2 | XP_011533240.1 | 6 - 7 | 537 | 130 | |
XM_017020395.1 | XP_016875884.1 | 8 - 9 | 792 | 130 | |
GenBank mRNA | AB101481.1 | - | 7 - 8 | 589 | 130 |
AK291273.1 | - | 7 - 8 | 688 | 130 | |
AY190526.1 | - | 7 - 8 | 589 | 130 | |
BC047395.1 | - | 2 - 3 | 213 | 130 | |
BC068595.1 | - | 7 - 8 | 679 | 130 |