The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_015193.4 | NP_056008.1 | 1 - 2 | 1993 | 92 |
GenBank mRNA | AF193421.1 | - | 1 - 2 | 1981 | 92 |
BC012321.1 | - | 1 - 2 | 1997 | 92 | |
BC075802.1 | - | 1 - 2 | 1985 | 92 | |
D87468.1 | - | 1 - 2 | 1985 | 92 |