Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
SLC2A8
Entrez Gene ID
29988
Gene Name
solute carrier family 2 member 8
Gene Aliases
GLUT8, GLUTX1
Chromosome Location
Chr.9: 127397138 - 127407898 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271711.1 NP_001258640.1 2 - 3 315 118
NM_014580.4 NP_055395.2 2 - 3 315 118
XM_006717084.3 XP_006717147.1 2 - 3 310 118
XM_011518602.2 XP_011516904.1 2 - 3 309 118
GenBank mRNA AF289587.1 - 2 - 3 222 118
AJ245937.1 - 2 - 3 225 118
BC019043.2 - 2 - 3 267 118
Y17801.1 - 2 - 3 248 118

View Full Product Details