The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001271711.1 | NP_001258640.1 | 7 - 8 | 1066 | 77 |
NM_001271712.1 | NP_001258641.1 | 5 - 6 | 696 | 77 | |
NM_014580.4 | NP_055395.2 | 7 - 8 | 1066 | 77 | |
NR_073416.1 | 4 - 5 | 499 | 77 | ||
XM_006717084.3 | XP_006717147.1 | 7 - 8 | 1061 | 77 | |
XM_011518602.2 | XP_011516904.1 | 7 - 8 | 982 | 77 | |
XM_011518603.2 | XP_011516905.1 | 5 - 6 | 721 | 77 | |
XM_011518604.2 | XP_011516906.1 | 5 - 6 | 645 | 77 | |
XM_017014653.1 | XP_016870142.1 | 5 - 6 | 686 | 77 | |
GenBank mRNA | AF289587.1 | - | 7 - 8 | 974 | 77 |
AJ245937.1 | - | 7 - 8 | 976 | 77 | |
BC019043.2 | - | 7 - 8 | 1018 | 77 | |
BM044230.1 | - | 4 - 5 | 400 | 77 | |
Y17801.1 | - | 7 - 8 | 999 | 77 |