The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005692.4 | NP_005683.2 | 1 - 2 | 211 | 92 |
NM_007189.2 | NP_009120.1 | 1 - 2 | 211 | 92 | |
GenBank mRNA | AF261091.1 | - | 1 - 2 | 68 | 92 |
AF261092.1 | - | 1 - 2 | 49 | 92 | |
HY165500.1 | - | 1 - 2 | 215 | 92 |