Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
AK7
Entrez Gene ID
122481
Gene Name
adenylate kinase 7
Gene Aliases
AK 7
Chromosome Location
Chr.14: 96392111 - 96489427 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_152327.3 NP_689540.2 14 - 15 1600 85
XM_006720021.2 XP_006720084.1 14 - 15 1600 85
XM_006720022.2 XP_006720085.1 13 - 14 1522 85
XM_017020957.1 XP_016876446.1 13 - 14 1522 85
GenBank mRNA AK057426.1 - 14 - 15 1600 85
BC035256.1 - 14 - 15 1589 85

View Full Product Details