The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001319135.1 | NP_001306064.1 | 7 - 8 | 932 | 81 |
NM_001322929.1 | NP_001309858.1 | 7 - 8 | 1144 | 81 | |
NM_001322930.1 | NP_001309859.1 | 5 - 6 | 946 | 81 | |
NM_001322931.1 | NP_001309860.1 | 8 - 9 | 1124 | 81 | |
NM_004602.3 | NP_004593.2 | 6 - 7 | 855 | 81 | |
NM_017452.3 | NP_059346.2 | 5 - 6 | 732 | 81 | |
NM_017453.3 | NP_059347.2 | 6 - 7 | 1021 | 81 | |
NM_017454.3 | NP_059348.2 | 4 - 5 | 657 | 81 | |
XM_017028028.1 | XP_016883517.1 | 7 - 8 | 998 | 81 | |
GenBank mRNA | AB209561.1 | - | 6 - 7 | 886 | 81 |
AF061938.1 | - | 5 - 6 | 726 | 81 | |
AF061939.1 | - | 6 - 7 | 1015 | 81 | |
AF061940.1 | - | 4 - 5 | 651 | 81 | |
AF061941.1 | - | 6 - 7 | 858 | 81 | |
AJ132258.1 | - | 3 - 4 | 399 | 81 | |
AK025519.1 | - | 5 - 6 | 695 | 81 | |
AK057739.1 | - | 6 - 7 | 978 | 81 | |
AK291487.1 | - | 6 - 7 | 936 | 81 | |
AK292859.1 | - | 5 - 6 | 517 | 81 | |
AL136601.2 | - | 5 - 6 | 715 | 81 | |
BC050432.1 | - | 5 - 6 | 759 | 81 | |
BC095397.1 | - | 5 - 6 | 704 | 81 |