The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001178098.1 | NP_001171569.1 | 13 - 14 | 1650 | 62 |
NM_001770.5 | NP_001761.3 | 13 - 14 | 1647 | 62 | |
XM_006721103.3 | XP_006721166.1 | 12 - 13 | 1587 | 62 | |
XM_017023893.1 | XP_016879382.1 | 12 - 13 | 1448 | 62 | |
GenBank mRNA | AA760729.1 | - | 2 - 3 | 62 | |
AK130657.1 | - | 12 - 13 | 1033 | 62 | |
AK301167.1 | - | 13 - 14 | 1565 | 62 | |
AK304456.1 | - | 14 - 15 | 1432 | 62 | |
AK313577.1 | - | 13 - 14 | 1621 | 62 | |
BC006338.2 | - | 13 - 14 | 1629 | 62 | |
M21097.1 | - | 13 - 14 | 1600 | 62 | |
X13312.1 | - | 13 - 14 | 1599 | 62 |