The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001206983.2 | NP_001193912.1 | 1 - 2 | 385 | 146 |
NM_001302703.1 | NP_001289632.1 | 2 - 3 | 295 | 146 | |
XM_006721676.3 | XP_006721739.1 | 1 - 2 | 329 | 146 | |
XM_017024145.1 | XP_016879634.1 | 2 - 3 | 262 | 146 | |
GenBank mRNA | BI819093.1 | - | 1 - 2 | 345 | 146 |
BX396749.2 | - | 2 - 3 | 282 | 146 | |
DA094787.1 | - | 1 - 2 | 378 | 146 |