The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001669.2 | NP_001001669.2 | 12 - 13 | 1886 | 70 |
XM_005268447.4 | XP_005268504.1 | 12 - 13 | 1857 | 70 | |
XM_005268448.1 | XP_005268505.1 | 11 - 12 | 1760 | 70 | |
XM_006714784.3 | XP_006714847.1 | 13 - 14 | 2225 | 70 | |
XM_011537642.2 | XP_011535944.1 | 12 - 13 | 1908 | 70 | |
XM_017009460.1 | XP_016864949.1 | 11 - 12 | 1999 | 70 | |
XM_017009461.1 | XP_016864950.1 | 10 - 11 | 1649 | 70 | |
GenBank mRNA | AB199796.1 | - | 11 - 12 | 70 | |
AK123597.1 | - | 4 - 5 | 70 | ||
AK300355.1 | - | 11 - 12 | 70 | ||
AK300559.1 | - | 12 - 13 | 70 | ||
AK316002.1 | - | 12 - 13 | 70 | ||
BF510234.1 | - | 2 - 3 | 188 | 70 |