Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
IL34
Entrez Gene ID
146433
Gene Name
interleukin 34
Gene Aliases
C16orf77, IL-34
Chromosome Location
Chr.16: 70579895 - 70660682 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001172772.1 NP_001166243.1 4 - 5 780 83
NM_152456.2 NP_689669.2 4 - 5 797 83
XM_011522902.2 XP_011521204.1 4 - 5 881 83
XM_011522903.2 XP_011521205.1 4 - 5 663 83
GenBank mRNA AK172744.1 - 3 - 4 312 83
AK307727.1 - 3 - 4 432 83
AK314913.1 - 3 - 4 317 83
BC029804.1 - 3 - 4 625 83
EU636994.1 - 3 - 4 242 83

View Full Product Details