The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001132.1 | NP_001001132.1 | 17 - 18 | 2224 | 103 |
NM_003024.2 | NP_003015.2 | 17 - 18 | 2224 | 103 | |
XM_005261026.2 | XP_005261083.1 | 17 - 18 | 2302 | 103 | |
XM_005261027.1 | XP_005261084.1 | 17 - 18 | 2302 | 103 | |
XM_005261031.1 | XP_005261088.1 | 17 - 18 | 2302 | 103 | |
XM_011529691.1 | XP_011527993.1 | 17 - 18 | 2191 | 103 | |
XM_011529692.1 | XP_011527994.1 | 17 - 18 | 2191 | 103 | |
XM_011529693.2 | XP_011527995.1 | 17 - 18 | 2302 | 103 | |
XM_017028428.1 | XP_016883917.1 | 18 - 19 | 2222 | 103 | |
XM_017028429.1 | XP_016883918.1 | 17 - 18 | 2191 | 103 | |
XM_017028430.1 | XP_016883919.1 | 17 - 18 | 2191 | 103 | |
XM_017028431.1 | XP_016883920.1 | 17 - 18 | 2302 | 103 | |
XM_017028432.1 | XP_016883921.1 | 17 - 18 | 2302 | 103 | |
XM_017028433.1 | XP_016883922.1 | 17 - 18 | 2302 | 103 | |
XM_017028434.1 | XP_016883923.1 | 17 - 18 | 2302 | 103 | |
XM_017028435.1 | XP_016883924.1 | 17 - 18 | 2191 | 103 | |
XM_017028436.1 | XP_016883925.1 | 17 - 18 | 2191 | 103 | |
XM_017028437.1 | XP_016883926.1 | 12 - 13 | 1642 | 103 | |
XM_017028438.1 | XP_016883927.1 | 17 - 18 | 2191 | 103 | |
XM_017028439.1 | XP_016883928.1 | 17 - 18 | 2302 | 103 | |
XM_017028440.1 | XP_016883929.1 | 17 - 18 | 2191 | 103 | |
XM_017028441.1 | XP_016883930.1 | 17 - 18 | 2302 | 103 | |
GenBank mRNA | AF064243.1 | - | 17 - 18 | 2062 | 103 |
AF064244.1 | - | 17 - 18 | 2062 | 103 | |
AF114487.1 | - | 17 - 18 | 2224 | 103 | |
AF114488.1 | - | 17 - 18 | 2224 | 103 | |
AK300274.1 | - | 17 - 18 | 2199 | 103 | |
BC116185.1 | - | 17 - 18 | 2035 | 103 | |
BC116186.2 | - | 17 - 18 | 2035 | 103 | |
BC117560.1 | - | 17 - 18 | 2204 | 103 | |
BX538175.1 | - | 12 - 13 | 1523 | 103 | |
DQ340865.1 | - | 16 - 17 | 1952 | 103 | |
DQ386455.1 | - | 16 - 17 | 1975 | 103 | |
DQ679754.1 | - | 17 - 18 | 2041 | 103 | |
DQ679755.1 | - | 17 - 18 | 2041 | 103 | |
DQ679756.1 | - | 18 - 19 | 1930 | 103 | |
DQ679757.1 | - | 17 - 18 | 2041 | 103 | |
EU107386.1 | - | 17 - 18 | 1930 | 103 | |
EU117382.1 | - | 16 - 17 | 1957 | 103 | |
EU120733.1 | - | 16 - 17 | 1957 | 103 | |
EU120734.1 | - | 16 - 17 | 1957 | 103 | |
EU120735.1 | - | 17 - 18 | 1930 | 103 | |
EU120736.1 | - | 17 - 18 | 1930 | 103 | |
EU140799.1 | - | 17 - 18 | 2041 | 103 | |
EU140800.1 | - | 16 - 17 | 1846 | 103 | |
EU152331.1 | - | 16 - 17 | 1957 | 103 | |
EU191901.1 | - | 16 - 17 | 1880 | 103 | |
GU939616.1 | - | 12 - 13 | 1646 | 103 | |
GU939617.1 | - | 12 - 13 | 1668 | 103 | |
U61166.1 | - | 1 - 2 | 120 | 103 |