The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_057749.2 | NP_477097.1 | 10 - 11 | 1045 | 131 |
XM_017013957.1 | XP_016869446.1 | 9 - 10 | 1370 | 131 | |
XM_017013958.1 | XP_016869447.1 | 10 - 11 | 1187 | 131 | |
XM_017013959.1 | XP_016869448.1 | 10 - 11 | 1289 | 131 | |
GenBank mRNA | AF091433.1 | - | 9 - 10 | 978 | 131 |
AF102778.1 | - | 8 - 9 | 941 | 131 | |
AF106690.1 | - | 8 - 9 | 941 | 131 | |
AF112857.1 | - | 8 - 9 | 806 | 131 | |
BC007015.1 | - | 8 - 9 | 780 | 131 | |
BC020729.1 | - | 10 - 11 | 992 | 131 |