Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ZDHHC4
Entrez Gene ID
55146
Gene Name
zinc finger DHHC-type containing 4
Gene Aliases
ZNF374
Chromosome Location
Chr.7: 6577413 - 6588996 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134387.1 NP_001127859.1 3 - 4 562 133
NM_001134388.1 NP_001127860.1 4 - 5 471 133
NM_001134389.1 NP_001127861.1 3 - 4 340 133
NM_018106.3 NP_060576.1 3 - 4 405 133
XM_005249793.2 XP_005249850.1 4 - 5 515 133
XM_005249794.3 XP_005249851.1 3 - 4 505 133
XM_005249795.2 XP_005249852.1 3 - 4 338 133
XM_005249796.3 XP_005249853.1 3 - 4 452 133
GenBank mRNA AF201931.1 - 2 - 3 133
AK001341.1 - 3 - 4 321 133
AK223532.1 - 3 - 4 405 133
AK308971.1 - 3 - 4 320 133
AL136674.1 - 3 - 4 313 133
AY359090.1 - 3 - 4 493 133
BC001239.1 - 4 - 5 424 133
BQ925460.1 - 4 - 5 431 133
CN365375.1 - 3 - 4 521 134
CR533511.1 - 1 - 2 113 133

View Full Product Details