Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ZDHHC4
Entrez Gene ID
55146
Gene Name
zinc finger DHHC-type containing 4
Gene Aliases
ZNF374
Chromosome Location
Chr.7: 6577413 - 6588996 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001134388.1 NP_001127860.1 1 - 2 65 151

View Full Product Details