Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- AKAP6
- Entrez Gene ID
-
9472
- Gene Name
- A-kinase anchoring protein 6
- Gene Aliases
-
ADAP100, ADAP6, AKAP100, PRKA6, mAKAP
- Chromosome Location
-
Chr.14: 32329273 - 32833999 on Build GRCh38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK094160.1 |
- |
1 - 2 |
805 |
104 |
View Full Product Details