Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TSHR
Entrez Gene ID
7253
Gene Name
thyroid stimulating hormone receptor
Gene Aliases
CHNG1, LGR3, hTSHR-I
Chromosome Location
Chr.14: 80954989 - 81146302 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000369.2 NP_000360.2 2 - 3 393 86
NM_001018036.2 NP_001018046.1 2 - 3 393 86
NM_001142626.2 NP_001136098.1 2 - 3 393 86
XM_005268037.4 XP_005268094.1 3 - 4 613 86
XM_005268039.1 XP_005268096.1 3 - 4 613 86
XM_006720245.1 XP_006720308.1 3 - 4 613 86
XM_011537119.2 XP_011535421.1 2 - 3 289 86
GenBank mRNA AB209207.1 - 2 - 3 265 86
AY429111.1 - 2 - 3 237 86
BC009237.2 - 2 - 3 302 86
BC024205.2 - 2 - 3 487 86
BC063613.1 - 2 - 3 269 86
BC108653.1 - 2 - 3 310 86
BC120972.2 - 2 - 3 263 86
BC120973.2 - 2 - 3 263 86
BC127628.1 - 2 - 3 263 86
BC141970.1 - 2 - 3 263 86
M31774.1 - 2 - 3 337 86
M32215.1 - 2 - 3 281 86
M73747.1 - 2 - 3 307 86
S45272.1 - 2 - 3 237 86
S49816.1 - 2 - 3 258 86

View Full Product Details