Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CNBD1
Entrez Gene ID
168975
Gene Name
cyclic nucleotide binding domain containing 1
Gene Aliases
-
Chromosome Location
Chr.8: 86866442 - 87428652 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_173538.2 NP_775809.1 1 - 2 130 119
XM_017013149.1 XP_016868638.1 1 - 2 136 119
GenBank mRNA AK093121.1 - 1 - 2 133 119
BC117326.1 - 1 - 2 130 119
BC117328.1 - 1 - 2 130 119

View Full Product Details