The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_006703.3 | NP_006694.1 | 1 - 2 | 420 | 73 |
GenBank mRNA | AF062529.1 | - | 1 - 2 | 255 | 73 |
AF062530.1 | - | 1 - 2 | 255 | 73 | |
AK313440.1 | - | 1 - 2 | 323 | 73 | |
BC007727.2 | - | 1 - 2 | 381 | 73 | |
BG714472.1 | - | 1 - 2 | 419 | 73 | |
BT019984.1 | - | 1 - 2 | 102 | 73 | |
BT019985.1 | - | 1 - 2 | 102 | 73 | |
DA803962.1 | - | 1 - 2 | 420 | 73 |