The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001261834.1 | NP_001248763.1 | 5 - 6 | 1179 | 58 |
NM_004749.3 | NP_004740.2 | 5 - 6 | 1223 | 58 | |
GenBank mRNA | AF011792.1 | - | 3 - 4 | 561 | 58 |
AK092132.1 | - | 3 - 4 | 2357 | 58 | |
AK298155.1 | - | 3 - 4 | 572 | 58 | |
AK300489.1 | - | 5 - 6 | 1138 | 58 | |
AL833840.1 | - | 5 - 6 | 1192 | 58 | |
BC002732.2 | - | 3 - 4 | 662 | 58 | |
BC014918.1 | - | 5 - 6 | 1153 | 58 | |
BC017235.2 | - | 5 - 6 | 1150 | 58 |