The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289905.1 | NP_001276834.1 | 5 - 6 | 679 | 69 |
NM_014339.6 | NP_055154.3 | 5 - 6 | 679 | 69 | |
GenBank mRNA | BC011624.2 | - | 5 - 6 | 623 | 69 |
BX334547.2 | - | 5 - 6 | 682 | 69 | |
BX410574.2 | - | 5 - 6 | 595 | 69 | |
CT841520.1 | - | 5 - 6 | 602 | 69 | |
JF305973.1 | - | 5 - 6 | 546 | 69 | |
U58917.1 | - | 5 - 6 | 578 | 69 |