The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001289905.1 | NP_001276834.1 | 7 - 8 | 895 | 58 |
NM_014339.6 | NP_055154.3 | 7 - 8 | 895 | 58 | |
GenBank mRNA | BC011624.2 | - | 7 - 8 | 839 | 58 |
BX368715.2 | - | 1 - 2 | 66 | 58 | |
BX410574.2 | - | 7 - 8 | 811 | 58 | |
CT841520.1 | - | 7 - 8 | 818 | 58 | |
JF305973.1 | - | 7 - 8 | 762 | 58 | |
U58917.1 | - | 7 - 8 | 794 | 58 |