The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001011544.1 | NP_001011544.1 | 4 - 5 | 370 | 72 |
NM_005366.4 | NP_005357.2 | 4 - 5 | 369 | 72 | |
XM_011531164.2 | XP_011529466.1 | 3 - 4 | 1000 | 72 | |
XM_017029521.1 | XP_016885010.1 | 6 - 7 | 580 | 72 | |
XM_017029522.1 | XP_016885011.1 | 6 - 7 | 1964 | 72 | |
GenBank mRNA | AK127849.1 | - | 6 - 7 | 1857 | 72 |
AY747607.1 | - | 4 - 5 | 327 | 72 | |
BC004479.2 | - | 4 - 5 | 305 | 72 | |
BQ212898.1 | - | 4 - 5 | 369 | 72 | |
BX337921.2 | - | 4 - 5 | 379 | 72 |