Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
FRMD7
Entrez Gene ID
90167
Gene Name
FERM domain containing 7
Gene Aliases
NYS, NYS1, XIPAN
Chromosome Location
Chr.X: 132076986 - 132128022 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306193.1 NP_001293122.1 11 - 12 1180 68
NM_194277.2 NP_919253.1 11 - 12 1227 68
XM_017029947.1 XP_016885436.1 11 - 12 1406 68
XM_017029948.1 XP_016885437.1 8 - 9 868 68
XM_017029949.1 XP_016885438.1 7 - 8 1189 68
GenBank mRNA AK125336.1 - 11 - 12 1227 68
AL161984.1 - 7 - 8 889 68
BC114371.1 - 11 - 12 1194 68
FJ717411.1 - 11 - 12 1004 68

View Full Product Details