Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CHSY1
Entrez Gene ID
22856
Gene Name
chondroitin sulfate synthase 1
Gene Aliases
CHSY, CSS1, ChSy-1, TPBS
Chromosome Location
Chr.15: 101175723 - 101251932 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_014918.4 NP_055733.2 2 - 3 1298 123
GenBank mRNA AB023207.1 - 2 - 3 1316 123
AB071402.1 - 2 - 3 836 123
BC046247.1 - 2 - 3 1305 123

View Full Product Details