The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_005502.3 | NP_005493.2 | 24 - 25 | 3932 | 67 |
XM_011518341.2 | XP_011516643.1 | 25 - 26 | 3916 | 67 | |
GenBank mRNA | AB055982.1 | - | 23 - 24 | 3528 | 67 |
AB445477.1 | - | 23 - 24 | 3528 | 67 | |
AF165281.1 | - | 22 - 23 | 3468 | 67 | |
AF285167.1 | - | 24 - 25 | 3818 | 67 | |
AJ012376.1 | - | 22 - 23 | 3468 | 67 | |
BC146856.1 | - | 24 - 25 | 3742 | 67 |