Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
CHD1
Entrez Gene ID
1105
Gene Name
chromodomain helicase DNA binding protein 1
Gene Aliases
-
Chromosome Location
Chr.5: 98855204 - 98929014 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001270.2 NP_001261.2 19 - 20 3008 141
XM_005271866.4 XP_005271923.1 20 - 21 3484 141
XM_005271867.4 XP_005271924.1 20 - 21 3484 141
XM_011543112.2 XP_011541414.1 20 - 21 3484 141
XM_017008991.1 XP_016864480.1 4 - 5 544 141
XM_017008992.1 XP_016864481.1 4 - 5 389 141
GenBank mRNA AF006513.1 - 19 - 20 3023 141
BC117134.1 - 19 - 20 2998 141
CN277742.1 - 2 - 3 173 141

View Full Product Details