The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001324281.1 | NP_001311210.1 | 21 - 22 | 3166 | 77 |
NM_032415.5 | NP_115791.3 | 20 - 21 | 3105 | 77 | |
XM_011515586.2 | XP_011513888.1 | 21 - 22 | 3134 | 77 | |
XM_011515587.2 | XP_011513889.1 | 20 - 21 | 3097 | 77 | |
GenBank mRNA | AF322641.1 | - | 20 - 21 | 3007 | 77 |
AF352576.1 | - | 18 - 19 | 2680 | 77 | |
AK074049.1 | - | 20 - 21 | 3025 | 77 | |
BC111719.1 | - | 19 - 20 | 2791 | 77 |