The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_024831.7 | NP_079107.6 | 12 - 13 | 2832 | 150 |
XM_005251328.2 | XP_005251385.1 | 11 - 12 | 2547 | 150 | |
GenBank mRNA | AF286340.1 | - | 12 - 13 | 2922 | 150 |
AF432215.1 | - | 12 - 13 | 2580 | 150 | |
AK026648.1 | - | 4 - 5 | 609 | 150 | |
AK055178.1 | - | 9 - 10 | 1706 | 150 | |
AK314693.1 | - | 12 - 13 | 2606 | 150 | |
AL832426.1 | - | 6 - 7 | 1088 | 150 | |
AY028423.1 | - | 12 - 13 | 2613 | 150 | |
AY534911.1 | - | 9 - 10 | 1622 | 150 | |
BC011999.1 | - | 5 - 6 | 726 | 150 |