Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
TRAPPC9
Entrez Gene ID
83696
Gene Name
trafficking protein particle complex 9
Gene Aliases
IBP, IKBKBBP, MRT13, NIBP, T1, TRS120
Chromosome Location
Chr.8: 139727725 - 140458579 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001160372.3 NP_001153844.1 10 - 11 1755 67
NM_001321646.1 NP_001308575.1 9 - 10 1728 67
NM_031466.7 NP_113654.4 10 - 11 1930 67
XM_011517326.2 XP_011515628.1 9 - 10 1903 67
XM_011517328.2 XP_011515630.1 10 - 11 1930 67
XM_011517330.2 XP_011515632.1 2 - 3 295 67
XM_017013893.1 XP_016869382.1 10 - 11 1930 67
XM_017013894.1 XP_016869383.1 3 - 4 631 67
GenBank mRNA AB067469.1 - 10 - 11 1705 67
AK026394.1 - 10 - 11 1747 67
AL833973.1 - 8 - 9 1030 67
AY190606.1 - 8 - 9 1009 67
BC006206.2 - 8 - 9 1151 67

View Full Product Details