The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138342.3 | NP_612351.2 | 18 - 19 | 2008 | 108 |
XM_006718937.1 | XP_006719000.1 | 18 - 19 | 1948 | 108 | |
GenBank mRNA | AF173889.1 | - | 17 - 18 | 1635 | 108 |
AK074703.1 | - | 18 - 19 | 1893 | 108 | |
AK074750.1 | - | 9 - 10 | 844 | 108 | |
AK074855.1 | - | 18 - 19 | 1878 | 108 | |
AK074947.1 | - | 18 - 19 | 1832 | 108 | |
AY358497.1 | - | 18 - 19 | 1888 | 108 | |
BC008326.1 | - | 14 - 15 | 1295 | 108 | |
BC040641.1 | - | 18 - 19 | 1970 | 108 | |
BX640934.1 | - | 19 - 20 | 4151 | 108 |