The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016234.3 | NP_057318.2 | 1 - 2 | 256 | 141 |
XM_017016324.1 | XP_016871813.1 | 1 - 2 | 291 | 141 | |
GenBank mRNA | AB033899.1 | - | 1 - 2 | 130 | 141 |
AK024932.1 | - | 1 - 2 | 257 | 141 | |
AK222782.1 | - | 1 - 2 | 274 | 141 | |
AK223288.1 | - | 1 - 2 | 245 | 141 | |
AL708480.1 | - | 1 - 2 | 257 | 141 | |
AY358520.1 | - | 1 - 2 | 217 | 141 | |
BC007985.2 | - | 1 - 2 | 234 | 141 |