Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
ACSL5
Entrez Gene ID
51703
Gene Name
acyl-CoA synthetase long-chain family member 5
Gene Aliases
ACS2, ACS5, FACL5
Chromosome Location
Chr.10: 112374158 - 112428380 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016234.3 NP_057318.2 6 - 7 819 86
NM_203379.1 NP_976313.1 6 - 7 680 86
NM_203380.1 NP_976314.1 6 - 7 846 86
XM_017016324.1 XP_016871813.1 6 - 7 854 86
GenBank mRNA AB033899.1 - 6 - 7 693 86
AK000339.1 - 6 - 7 829 86
AK024932.1 - 6 - 7 820 86
AK222782.1 - 6 - 7 837 86
AK223288.1 - 6 - 7 808 86
AK310436.1 - 6 - 7 629 86
AM262166.1 - 5 - 6 539 86
AY358520.1 - 6 - 7 780 86
BC007985.2 - 6 - 7 797 86

View Full Product Details