The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001172774.1 | NP_001166245.1 | 12 - 13 | 1520 | 91 |
NM_207325.2 | NP_997208.2 | 12 - 13 | 1546 | 91 | |
XM_006723045.1 | XP_006723108.1 | 9 - 10 | 1062 | 91 | |
XM_011526526.2 | XP_011524828.1 | 12 - 13 | 1501 | 91 | |
XM_017026366.1 | XP_016881855.1 | 11 - 12 | 1435 | 91 | |
XM_017026367.1 | XP_016881856.1 | 12 - 13 | 1460 | 91 | |
GenBank mRNA | AK126264.1 | - | 6 - 7 | 621 | 91 |
AK126757.1 | - | 12 - 13 | 1537 | 91 | |
AK129497.1 | - | 12 - 13 | 1501 | 91 | |
AL833713.1 | - | 2 - 3 | 242 | 91 | |
BC131725.1 | - | 11 - 12 | 1368 | 91 | |
CR749459.1 | - | 6 - 7 | 716 | 91 |