The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001398.2 | NP_001389.2 | 7 - 8 | 745 | 63 |
XM_017026448.1 | XP_016881937.1 | 7 - 8 | 695 | 63 | |
GenBank mRNA | AK126566.1 | - | 4 - 5 | 63 | |
AK291860.1 | - | 7 - 8 | 680 | 63 | |
BC011792.2 | - | 7 - 8 | 662 | 63 | |
BC014786.1 | - | 3 - 4 | 63 | ||
BC017408.1 | - | 7 - 8 | 686 | 63 |