Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
CD3E
Entrez Gene ID
916
Gene Name
CD3e molecule
Gene Aliases
IMD18, T3E, TCRE
Chromosome Location
Chr.11: 118304580 - 118316175 on Build GRCh38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_000733.3 NP_000724.1 8 - 9 825 108
GenBank mRNA BC049847.1 - 7 - 8 637 108
X03884.1 - 7 - 8 623 108

View Full Product Details