The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_194441.2 | NP_919423.1 | 1 - 2 | 175 | 74 |
GenBank mRNA | AK291162.1 | - | 1 - 2 | 175 | 74 |
T86577.1 | - | 1 - 2 | 68 | 74 |