The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001318860.1 | NP_001305789.1 | 4 - 5 | 679 | 74 |
NM_001318861.1 | NP_001305790.1 | 5 - 6 | 765 | 74 | |
NM_001318862.1 | NP_001305791.1 | 5 - 6 | 747 | 74 | |
NM_001318863.1 | NP_001305792.1 | 4 - 5 | 661 | 74 | |
NM_080916.2 | NP_550438.1 | 5 - 6 | 792 | 74 | |
NR_134898.1 | 3 - 4 | 531 | 74 | ||
XM_011532647.2 | XP_011530949.1 | 5 - 6 | 770 | 74 | |
GenBank mRNA | AI201630.1 | - | 2 - 3 | 74 | |
BC015757.1 | - | 5 - 6 | 731 | 74 | |
BG470729.1 | - | 4 - 5 | 595 | 74 | |
BG575800.1 | - | 5 - 6 | 670 | 74 | |
BG708680.1 | - | 4 - 5 | 668 | 74 | |
CA414889.1 | - | 2 - 3 | 74 | ||
KU178023.1 | - | 5 - 6 | 707 | 74 | |
KU178024.1 | - | 4 - 5 | 594 | 74 | |
U41668.1 | - | 5 - 6 | 741 | 74 |