The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_020314.5 | NP_064710.4 | 14 - 15 | 1541 | 87 |
XM_005255435.3 | XP_005255492.1 | 14 - 15 | 1493 | 87 | |
XM_011545896.2 | XP_011544198.1 | 14 - 15 | 1242 | 87 | |
XM_011545897.2 | XP_011544199.1 | 7 - 8 | 609 | 87 | |
GenBank mRNA | AF461052.2 | - | 8 - 9 | 1037 | 87 |
AK024693.1 | - | 10 - 11 | 811 | 87 | |
AK290286.1 | - | 14 - 15 | 1238 | 87 | |
AL136744.1 | - | 1 - 2 | 154 | 87 | |
AL832159.1 | - | 14 - 15 | 1237 | 87 | |
BC014900.2 | - | 13 - 14 | 1209 | 87 | |
BC050464.1 | - | 14 - 15 | 1281 | 87 | |
BC058845.1 | - | 14 - 15 | 1240 | 87 | |
BX537867.1 | - | 14 - 15 | 1253 | 87 |